ENA408616649 Enamine

2-cyclopropyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD12026856
  • Cas Number : 1060816-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97481

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-97481
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart