ENA410542301 Enamine

1-(2-bromoethyl)-4-cyclopropylpiperazine hydrobromide

  • MDL number MFCD26407521
  • Cas Number : 1803607-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115515
Availability details for
Quantity
  • Availability
  • Add to Cart