ENA410542311 Enamine

N-[3-(piperidin-3-yl)-1H-pyrazol-4-yl]acetamide

  • MDL number MFCD26407554
  • Cas Number : 1803597-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129896
Availability details for
Quantity
  • Availability
  • Add to Cart