ENA410542312 Enamine

N-[3-(piperidin-3-yl)-1H-pyrazol-4-yl]methanesulfonamide

  • MDL number MFCD26407555
  • Cas Number : 1803566-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129897
Availability details for
Quantity
  • Availability
  • Add to Cart