ENA410542313 Enamine

3-ethyl-1-[3-(piperidin-3-yl)-1H-pyrazol-4-yl]urea

  • MDL number MFCD26407556
  • Cas Number : 1803585-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129898
Availability details for
Quantity
  • Availability
  • Add to Cart