ENA410542314 Enamine

N-[3-(piperidin-3-yl)-1H-pyrazol-4-yl]cyclopentanecarboxamide

  • MDL number MFCD26407557
  • Cas Number : 1803598-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129901
Availability details for
Quantity
  • Availability
  • Add to Cart