ENA410542316 Enamine

3,3,3-trifluoro-2-methylpropan-1-amine hydrochloride

  • MDL number MFCD26407559
  • Cas Number : 2794-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 266-268
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130024
Availability details for
Quantity
  • Availability
  • Add to Cart