ENA410542326 Enamine

2-amino-3-(4-methylbenzoyl)-4H,5H,7H-1,6$l^{6}-thieno[2,3-c][1$l^{6}]thiopyran-6,6-dione

  • MDL number MFCD26407606
  • Cas Number : 1803583-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133857
Availability details for
Quantity
  • Availability
  • Add to Cart