ENA410542328 Enamine

6-bromo-8-methylimidazo[1,2-a]pyridine-2-carboxamide

  • MDL number MFCD26407621
  • Cas Number : 1803585-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134424
Availability details for
Quantity
  • Availability
  • Add to Cart