ENA410542334 Enamine

1,3-diethyl 2-(but-2-yn-1-yl)-2-acetamidopropanedioate

  • MDL number MFCD26407662
  • Cas Number : 19013-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135889
Availability details for
Quantity
  • Availability
  • Add to Cart