ENA410542335 Enamine

1,3-diethyl 2-(but-3-en-1-yl)-2-acetamidopropanedioate

  • MDL number MFCD26407731
  • Cas Number : 122471-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46...48
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136461
Availability details for
Quantity
  • Availability
  • Add to Cart