ENA410542336 Enamine

tert-butyl N-[1-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)hex-4-en-2-yl]carbamate

  • Cas Number : 1807941-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136509
Availability details for
Quantity
  • Availability
  • Add to Cart