ENA410542338 Enamine

2-[2-(1-methyl-1H-indol-3-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD26407768
  • Cas Number : 70369-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136829
Availability details for
Quantity
  • Availability
  • Add to Cart