ENA410542345 Enamine

3-{N-[(3-fluorophenyl)methyl]acetamido}propanoic acid

  • MDL number MFCD26407812
  • Cas Number : 1803611-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137171
Availability details for
Quantity
  • Availability
  • Add to Cart