ENA410542346 Enamine

4-[4-amino-2-(trifluoromethyl)phenyl]piperazine-1-carbaldehyde

  • MDL number MFCD26407831
  • Cas Number : 1803599-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137316
Availability details for
Quantity
  • Availability
  • Add to Cart