ENA410542347 Enamine

methyl 4-butylmorpholine-3-carboxylate

  • MDL number MFCD26407832
  • Cas Number : 1565318-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137334
Availability details for
Quantity
  • Availability
  • Add to Cart