ENA410542348 Enamine

methyl 4-(cyclobutylmethyl)morpholine-3-carboxylate

  • MDL number MFCD26407833
  • Cas Number : 1706584-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137337
Availability details for
Quantity
  • Availability
  • Add to Cart