ENA410542350 Enamine

2,2,2-trifluoro-N-(1-methyl-1H-pyrazol-5-yl)acetamide

  • MDL number MFCD26407842
  • Cas Number : 1693994-53-4
  • Molecular Weight : 193.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137408
Availability details for
Quantity
  • Availability
  • Add to Cart