ENA410542351 Enamine

1,3-dichloro-2-ethoxy-5-nitrobenzene

  • MDL number MFCD26407852
  • Cas Number : 62047-40-9
  • Molecular Weight : 236.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dichloro-2-ethoxy-5-nitrobenzene
Inchi Code 1S/C8H7Cl2NO3/c1-2-14-8-6(9)3-5(11(12)13)4-7(8)10/h3-4H,2H2,1H3
Inchi Key NRZAIWWKGAZYDY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137489
Availability details for
Quantity
  • Availability
  • Add to Cart