ENA410542352 Enamine

5-methyl-4-(2-nitrophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD26407857
  • Cas Number : 1803598-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137540
Availability details for
Quantity
  • Availability
  • Add to Cart