ENA410542353 Enamine

3-(2-nitroethyl)thiophene

  • MDL number MFCD26407863
  • Cas Number : 30807-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137587
Availability details for
Quantity
  • Availability
  • Add to Cart