ENA410542354 Enamine

2-chloro-4-(dichloromethyl)pyrimidine

  • MDL number MFCD26407864
  • Cas Number : 1803566-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137609
Availability details for
Quantity
  • Availability
  • Add to Cart