ENA410542362 Enamine

1-[4-(1,4-diazepan-1-yl)phenyl]ethan-1-one hydrochloride

  • MDL number MFCD26407898
  • Cas Number : 1803566-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137839
Availability details for
Quantity
  • Availability
  • Add to Cart