ENA410542364 Enamine

1-[(2-cyano-6-methylphenyl)sulfanyl]-N,N-dimethylformamide

  • MDL number MFCD26407902
  • Cas Number : 1803609-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137867
Availability details for
Quantity
  • Availability
  • Add to Cart