ENA410542367 Enamine

2-{[4-(1-benzofuran-2-yl)-1,3-thiazol-2-yl]methyl}-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD26407906
  • Cas Number : 1803571-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137910
Availability details for
Quantity
  • Availability
  • Add to Cart