ENA410542370 Enamine

methyl 1-(4-chlorophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD26407915
  • Cas Number : 1642802-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138046

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138046
Availability details for
Quantity
  • Availability
  • Add to Cart