ENA410542371 Enamine

tert-butyl 3-({[1,2,4]triazolo[4,3-a]pyridin-3-yl}methyl)piperidine-1-carboxylate

  • MDL number MFCD26407919
  • Cas Number : 1803608-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138209
Availability details for
Quantity
  • Availability
  • Add to Cart