ENA410542380 Enamine

(3-amino-2-benzylpropyl)benzene hydrochloride

  • MDL number MFCD26407932
  • Cas Number : 890715-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138306
Availability details for
Quantity
  • Availability
  • Add to Cart