ENA410542382 Enamine

5-oxo-1-{[2-(trifluoromethyl)phenyl]methyl}pyrrolidine-3-carboxamide

  • MDL number MFCD26407937
  • Cas Number : 1803572-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138357

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138357
Availability details for
Quantity
  • Availability
  • Add to Cart