ENA410542383 Enamine

(1R)-1-(3-methyl-1-benzothiophen-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD26407940
  • Cas Number : 1807940-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 87
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138589
Availability details for
Quantity
  • Availability
  • Add to Cart