ENA410542386 Enamine

2-[(2,2,2-trifluoroethyl)amino]acetamide hydrochloride

  • MDL number MFCD26407944
  • Cas Number : 1803592-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138676
Availability details for
Quantity
  • Availability
  • Add to Cart