ENA410542391 Enamine

sodium 2-{[4-(furan-2-ylmethyl)-5-(pyridin-3-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetate

  • MDL number MFCD26407955
  • Cas Number : 1803599-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138813
Availability details for
Quantity
  • Availability
  • Add to Cart