ENA410542393 Enamine

sodium 2-({4-[(furan-2-yl)methyl]-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl}sulfanyl)acetate

  • MDL number MFCD26407957
  • Cas Number : 1803598-64-2
  • Molecular Weight : 338.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138839
Availability details for
Quantity
  • Availability
  • Add to Cart