ENA410542395 Enamine

methyl 2-(4-methylpiperidin-1-yl)-2-oxoacetate

  • MDL number MFCD25978447
  • Cas Number : 1565657-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138932
Availability details for
Quantity
  • Availability
  • Add to Cart