ENA410542397 Enamine

2-(2-methyl-1-benzofuran-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD26407966
  • Cas Number : 1803584-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138945
Availability details for
Quantity
  • Availability
  • Add to Cart