ENA410542399 Enamine

3a,4,5,6,7,7a-hexahydro-1H-1,3-benzodiazol-2-amine hydrochloride

  • MDL number MFCD26407972
  • Cas Number : 91928-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139064

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139064
Availability details for
Quantity
  • Availability
  • Add to Cart