ENA410542411 Enamine

2-[2-(1,3-thiazol-2-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione hydrobromide

  • MDL number MFCD26407996
  • Cas Number : 1803587-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139302
Availability details for
Quantity
  • Availability
  • Add to Cart