ENA410542412 Enamine

1-(2-methoxyphenyl)-5-methyl-1,4-diazepane dihydrochloride

  • MDL number MFCD26407999
  • Cas Number : 1803599-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139367
Availability details for
Quantity
  • Availability
  • Add to Cart