ENA410542419 Enamine

2-[5-(methoxymethyl)furan-2-yl]pyrrolidine

  • MDL number MFCD26408008
  • Cas Number : 1596781-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139464

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139464
Availability details for
Quantity
  • Availability
  • Add to Cart