ENA410542421 Enamine

3-(methoxymethyl)azepane

  • MDL number MFCD26408011
  • Cas Number : 1566574-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139498
Availability details for
Quantity
  • Availability
  • Add to Cart