ENA410542423 Enamine

(2-aminoethyl)(2,2-difluoroethyl)methylamine hydrochloride

  • MDL number MFCD26408017
  • Cas Number : 1803583-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139584
Availability details for
Quantity
  • Availability
  • Add to Cart