ENA410542430 Enamine

2-(3,4-difluorophenyl)-2-methylcyclopropan-1-amine hydrochloride

  • MDL number MFCD26408025
  • Cas Number : 1803598-88-0
  • Molecular Weight : 219.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,4-difluorophenyl)-2-methylcyclopropan-1-amine hydrochloride
Inchi Code 1S/C10H11F2N.ClH/c1-10(5-9(10)13)6-2-3-7(11)8(12)4-6;/h2-4,9H,5,13H2,1H3;1H
Inchi Key BRNFGVSIVWTWAF-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-139730

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-139730
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart