ENA410542431 Enamine

4-(3-bromopropyl)isoquinoline hydrobromide

  • MDL number MFCD26408027
  • Cas Number : 1803592-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139753
Availability details for
Quantity
  • Availability
  • Add to Cart