ENA410542433 Enamine

(2-methanesulfonyl-6-methoxyphenyl)hydrazine hydrochloride

  • MDL number MFCD26408029
  • Cas Number : 1803598-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139793
Availability details for
Quantity
  • Availability
  • Add to Cart