ENA410542434 Enamine

4-(1H-1,2,3-triazol-4-yl)piperidine dihydrochloride

  • MDL number MFCD26408031
  • Cas Number : 1803606-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139858
Availability details for
Quantity
  • Availability
  • Add to Cart