ENA410542440 Enamine

4-methyl-1,2,3,4-tetrahydroquinolin-8-ol hydrobromide

  • MDL number MFCD26408039
  • Cas Number : 1803599-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139942
Availability details for
Quantity
  • Availability
  • Add to Cart