ENA410542441 Enamine

methyl 2-amino-2-(4-fluoro-3-methylphenyl)acetate hydrochloride

  • MDL number MFCD26408043
  • Cas Number : 1803566-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140009

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-140009
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart