ENA410542443 Enamine

2-(2,5-difluorophenyl)-2-methylcyclopropan-1-amine hydrochloride

  • MDL number MFCD26408050
  • Cas Number : 1803585-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140101
Availability details for
Quantity
  • Availability
  • Add to Cart