ENA410542446 Enamine

2-amino-N-ethyl-4-methyl-N-[(2-methylphenyl)methyl]pentanamide hydrochloride

  • MDL number MFCD26407523
  • Cas Number : 1427582-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117406
Availability details for
Quantity
  • Availability
  • Add to Cart