ENA410542447 Enamine

5-amino-1,3-dimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD18879234
  • Cas Number : 1342150-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117666
Availability details for
Quantity
  • Availability
  • Add to Cart